Learn More
Medchemexpress LLC Pulegone | 89-82-7 | 152.24 | 500 MG
SDP
Supplier:  Medchemexpress LLC HYN1500500MG
Encompass_Preferred
Pulegone is a monoterpene ketone compound commonly found in the essential oils of various plants. It acts as a bird repellent and demonstrates diverse biological activities, including anti-inflammatory, antibacterial, antifungal, and anti-hyperalgesic effects. It is notably effective against bacteria of the Salmonella species.
  • Inhibits expression of inflammatory mediators by downregulating iNOS, COX-2, NF-κB, and MAPK signaling pathways.
  • Upregulates Nrf-2/HO-1.
  • Increases intracellular Ca2+ concentration in chicken sensory neurons.
  • Activates TRPM8 and TRPA1.
  • Exhibits anti-hyperalgesic effects in inflammatory pain models.
  • Appears as a colorless to light yellow liquid.
  • Has a purity of 99.57%.
Catalog No. 50-004-16018