Learn More
Medchemexpress LLC Puliginurad (YL-90148) | 2013582-27-7 | 99.9% | 336.41 | 50 MG

Supplier: Medchemexpress LLC HY14730050MG
Puliginurad (YL-90148) is a potent urate transporter (URAT) inhibitor. It can be used for hyperuricemia and gout research.
- Shipping: room temperature in continental US; may vary elsewhere.
- General storage: 4°C, sealed storage, away from moisture and light.
- In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture and light).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.