Learn More
Analytical Sales & Services Pulse Damper - High Capacity
Supplier:  Analytical Sales & Services 73421
Encompass
High Capacity Pulse Damper. Features a bimodal damping mechanism. Increased damping efficiency over wider pressure range. Suited for damping at low pressures, < 500psi.\n\nQty/Pkg: EACH
Catalog No. 50-008-06502