Learn More
Medchemexpress LLC PUMA BH3 | 98.2% | 2384.63 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP15621MG
Encompass_Preferred
PUMA BH3 is a p53 upregulated modulator of apoptosis (PUMA) BH3 domain peptide, acting as a direct activator of Bak, with a Kd of 26 nM. It binds to Bak in the canonical BH3-binding groove and activates Bak.
  • Acts as a direct activator of Bak
  • Kd of 26 nM
  • Solid appearance
  • White to off-white color
  • Soluble in DMSO at 100 mg/mL
Catalog No. 50-004-22803