Learn More
Medchemexpress LLC PUMA BH3 | 98.2% | 2384.63 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP156210MG
Encompass_Preferred
PUMA BH3 is a p53 upregulated modulator of apoptosis (PUMA) BH3 domain peptide. It functions as a direct activator of Bak with a Kd of 26 nM. The peptide binds Bak in the canonical BH3-binding groove, leading to its activation.
  • P53 upregulated modulator of apoptosis (PUMA) BH3 domain peptide
  • Direct activator of Bak
  • Kd of 26 nM against Bak
  • Binds Bak in the canonical BH3-binding groove
Catalog No. 50-004-22802