Learn More
Cayman Chemical PUMA BH3humntrIfluoroac 5mg
SDP
Supplier:  Cayman Chemical 375095 MG
Encompass_Preferred
A peptide activator of Bak; binds to Bak (Kds = 290 and 26 nM in HEPES and CHAPS buffer, respectively) and induces Bak homo-oligomerization and Bak-mediated membrane permeabilization
Catalog No. 50-291-5686