Learn More
Medchemexpress LLC Pumecitinib | 2401057-12-1 | 99.8% | 400.46 | 5 MG

Supplier: Medchemexpress LLC HY1487915MG
Pumecitinib is a Janus kinase (JAK) inhibitor with anti-inflammatory activity, intended for research use only.
- Janus kinase (JAK) inhibitor
- Exhibits anti-inflammatory activity
- Chemical formula: C17H20N8O2S
- Appears as a white to off-white solid
- Soluble in DMSO with ultrasonic and warming
- Also soluble in 10% DMSO and 90% corn oil for in vivo studies
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.