Learn More
AVANTOR MASTERFLEX (VWR LLC) PUMP MFLX LS EL2 OH 115/230
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX7792179
Encompass
Masterflex L/S Digital Drive with Open-Head Sensor and Easy-Load II Pump Head for High-Performance Tubing, 600 rpm; 115/230 VAC
Catalog No. NC1917143