Learn More
Metrohm USA 843 VA Peristaltic Pump Station for Compact VA Systems
SDP
Supplier:  Metrohm USA 28430140
Encompass
The 843 Peristaltic Pump Station has two built-in peristaltic pumps that can be controlled directly by the 797 VA Computrace via the remote signal. It is used for the automatic rinsing and emptying of the measuring vessel in partly automated and fully automated VA and CVS systems with the 797 VA Computrace.
Catalog No. NC1808511