Learn More
Hach Company Pump Tubing, PVC 0.025 in (0.635 mm) I.D., Orange/White, 12-Pack - Replacement for Automated Lab Pumps and Flow Injection Analysis
Supplier:  Hach Company 53406
Encompass
PVC pump tubing with a 0.025 in (0.635 mm) interior diameter, supplied in an orange/white color and packaged as a 12-piece set. Designed for laboratory fluid handling, it is suitable for automated lab pumps and flow injection analysis (FIA) applications as replacement pump tubing.
  • Interior diameter: 0.025 in (0.635 mm).
  • Material: PVC.
  • Color aids visual identification and routing.
  • Packaged as 12 tubes for easy replacement and inventory stocking.
  • Suitable for automated lab pumps and flow injection analysis (FIA) systems.
Catalog No. NC3350993