Learn More
Essendant PUMPCHRM PLTDDRM6/CS
Supplier:  Essendant BWK00422
Encompass_Preferred
Corrosion-resistant chrome plating protects metal from the corrosive effects of water-based solutions One size fits most standard 15 30 and 55 gallon drums Large 22 oz output per stroke significantly reduces the number of times you pump resulting in less hand fatigue Broadly applicable pump is usable with a wide variety of solutions including water-based solutions and solvents Durable pump is ideal for thicker liquids like motor oil Do not use with corrosive chemicals Teflon coated packing seal increases chemical resistance Pump Application Type- Metal Drum Pump Style- Push Pump Operating Principle- Displacement Volume Number of Strokes- 22 oz 1
Catalog No. 50-000-23002