Learn More
Apexbio Technology LLC Punicalagin 65995-63-3 20mg
SDP
Supplier:  Apexbio Technology LLC N168720
Encompass_Preferred
Punicalagin (CAS 65995-63-3) is a small-molecule inhibitor targeting NF- B and MAPK signaling pathways It is designed to modulate these key inflammatory signaling cascades thereby regulating immune and inflammatory responses Punicalagin exerts its biological activity primarily through inhibition of inflammatory cytokine production and immune response signaling In cellular and animal-based studies punicalagin demonstrates inhibitory activity with IC50 values ranging from approximately 10 to 50 M depending on cell type and assay conditions Based on these pharmacological properties punicalagin holds research potential in immunology virology autoimmune disease models and inflammation-related pathogenesis
Catalog No. 50-000-89159