Learn More
Invivogen pUNO1 bearing the human PTPN22 gene, isoform 1, 20ug
SDP
Supplier:  Invivogen PUNO1HPTPN22
Encompass
Human PTPN22 isoform 1 ('LyP1') (pUNO1-hPTPN22a)Genbank : NM_015967.5ORF size : 2424 bpSubclone : AgeI - NheI
Catalog No. NC3732564