Learn More
Air Science Usa Air Science ASVLFP536-030 HEPA Filter, 99.99% Efficiency, Easy Tool-Free Replacement, 1 Unit
SDP
Supplier:  Air Science Usa ASVLFP536030
Encompass_Preferred
The Air Science USA ASVLFP536-030 is a HEPA filter designed for Purair PCR-36 and FLOW-36 laminar flow cabinets. With 99.99% efficiency in removing particles as small as 0.3 µm, it ensures a contaminant-free environment critical for PCR applications. Its lightweight aluminum frame allows for tool-free installation, making replacement easy and quick. This filter guarantees effective protection against particulate contamination, setting it apart from others in its category.
Catalog No. NC1969082