Learn More
Biolegend Purified anti-DYKDDDDK Tag
Supplier:  Biolegend 637303
Encompass_Preferred
Purified anti-DYKDDDDK Tag [L5]; Isotype: Rat IgG2a, λ; Reactivity: DYKDDDDK tag epitope; Apps: WB, IP, ICC, ELISA, FC, Purification; Size: 1 mg
Catalog No. 50-170-918