Learn More
Biolegend Purified anti-β-Dystroglycan
Supplier:  Biolegend 849401
Encompass_Preferred
Purified anti-β-Dystroglycan [4F7]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Mouse; Apps: WB, IHC-P, IP; Size: 25 ug
Catalog No. 50-207-4559