Learn More
Biolegend Purified anti-human CD303 (BDCA-2) (Maxpar Ready)
Supplier:  Biolegend 354215
Encompass_Preferred
Purified anti-human CD303 (BDCA-2) (Maxpar Ready) [201A]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC, CyTOF; Size: 100 ug
Catalog No. 50-207-7329