Learn More
Biolegend Purified anti-human Basophils (2D7)
Supplier:  Biolegend 346202
Encompass_Preferred
Purified anti-human Basophils (2D7) [Basophil/2D7]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: ICFC, ICC, IHC-P, IHC-F; Size: 100 ug
Catalog No. 50-168-368