Learn More
Biolegend Purified anti-human CD134 (OX40)
Supplier:  Biolegend 350002
Encompass_Preferred
Purified anti-human CD134 (OX40) [Ber-ACT35 (ACT35)]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee; Apps: FC, IHC-P, ELISA, IHC-F, WB; Size: 100 ug
Catalog No. 50-168-560