Learn More
Biolegend Purified anti-human CD235a (Glycophorin A)
Supplier:  Biolegend 349102
Encompass_Preferred
Purified anti-human CD235a (Glycophorin A) [HI264]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 100 ug
Catalog No. 50-168-505