Learn More
Biolegend Purified anti-human CD272 (BTLA)
Supplier:  Biolegend 344502
Encompass_Preferred
Purified anti-human CD272 (BTLA) [MIH26]; Isotype: Mouse IgG2a, κ; Reactivity: Human, African Green; Apps: FC, Agonist; Size: 100 ug
Catalog No. 50-168-247