Learn More
Biolegend Purified anti-human CD34
Supplier:  Biolegend 343502
Encompass_Preferred
Purified anti-human CD34 [581]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Cynomolgus; Apps: FC, CyTOF, ICC, IHC-P; Size: 100 ug
Catalog No. 50-168-183