Learn More
Biolegend Purified anti-human Galectin-9
Supplier:  Biolegend 348902
Encompass_Preferred
Purified anti-human Galectin-9 [9M1-3]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus; Apps: ICFC, IHC-P, FC, Block; Size: 100 ug
Catalog No. 50-168-496