Learn More
Biolegend Purified anti-human Siglec-9
Supplier:  Biolegend 351502
Encompass_Preferred
Purified anti-human Siglec-9 [K8]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC, ELISA, ICC, IP, WB; Size: 100 ug
Catalog No. 50-168-663