Learn More
Biolegend Purified anti-Mcl-1
Supplier:  Biolegend 695702
Encompass_Preferred
Purified anti-Mcl-1 [W16014A]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: WB, ICC; Size: 100 ug
Catalog No. 50-208-7665