Learn More
Biolegend Purified anti-mouse CD129 (IL-9R)
Supplier:  Biolegend 158702
Encompass_Preferred
Purified anti-mouse CD129 (IL-9R) [S18011E]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 500 ug
Catalog No. 50-207-2402