Learn More
Biolegend Purified anti-mouse CD138 (Syndecan-1)
Supplier:  Biolegend 142501
Encompass_Preferred
Purified anti-mouse CD138 (Syndecan-1) [281-2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, ICC, IHC-P, IHC-F; Size: 25 ug
Catalog No. 50-165-264