Learn More
Biolegend Purified anti-mouse CD2
Supplier:  Biolegend 100102
Encompass_Preferred
Purified anti-mouse CD2 [RM2-5]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC, IP, Block, Costim; Size: 500 ug
Catalog No. 50-162-360