Learn More
Biolegend Purified anti-mouse CD3ε
Supplier: Biolegend 100302
Purified anti-mouse CD3ε [145-2C11]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC, IHC-F, IP, Activ, Block, WB, ICC; Size: 500 ug