Learn More
Biolegend Purified anti-mouse Ly-51
Supplier:  Biolegend 108302
Encompass_Preferred
Purified anti-mouse Ly-51 [6C3]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC, IHC, IP; Size: 500 ug
Catalog No. 50-163-235