Learn More
Biolegend Purified anti-mouse TER-119/Erythroid Cells
Supplier:  Biolegend 116202
Encompass_Preferred
Purified anti-mouse TER-119/Erythroid Cells [TER-119]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC, IHC-F, IHC-P, IP, WB; Size: 500 ug
Catalog No. 50-163-619