Learn More
Biolegend Purified anti-mouse VEGFR-3 (FLT-4)
Supplier:  Biolegend 140901
Encompass_Preferred
Purified anti-mouse VEGFR-3 (FLT-4) [AFL4]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: WB, IHC-F, Block, ELISA, IP; Size: 50 ug
Catalog No. 50-165-166