Learn More
Biolegend Purified anti-PER1
Supplier:  Biolegend 936002
Encompass_Preferred
Purified anti-PER1 [W17208B]; Isotype: Rat IgG2a, κ; Reactivity: Human, Mouse; Apps: WB, IP, KO/KD-WB; Size: 100 ug
Catalog No. 50-207-5348