Learn More
Aqua Solutions Pure Water™ pH Buffers and pHISA™ Adjustor, Pack of 4
SDP
Supplier:  Aqua Solutions OR700702
Encompass
Sample well water, rain water, distilled/deionized water, boiler feed water, process water and more with the Thermo Scientific™ Orion™ Pure Water™ pH Buffers and pHISA™ Adjustor. It offers fast, accurate pH results in high purity and low ionic strength waters.
Catalog No. NC1768531