Learn More
CUBE BIOTECH Co-NTA Cartridge for His-tag
Supplier:  CUBE BIOTECH 31602
Encompass
1ml of Co-NTA Agarose slurry (50%) in a 1ml for Protein Purification of His-tagged proteins via FPLC
Catalog No. NC2684635