Learn More
CUBE BIOTECH PureCube Ni-IDA Agarose; 10 ml
Supplier:  CUBE BIOTECH 30103
Encompass
10ml of Ni-IDA His-tag Agarose slurry (50%) in a 1ml for Protein Purification of His-tagged proteins
Catalog No. NC2684618