Learn More
CUBE BIOTECH PureCube Ni-IDA MagBeads; 5 ml
Supplier:  CUBE BIOTECH 30205
Encompass
5ml of Ni-IDA His-tag MagBead slurry (50%) in a 1ml for Protein Purification of His-tagged proteins
Catalog No. NC2684621