Learn More
Bellco Glass, Inc. PUREGRIP Universal HPLC Solvent Reservoir, 38-430 Cap Assembly, 3 x Ports- OD Tube-3.2 mm (1/8") & 1.6 mm (1/16"), 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800065
Encompass_Preferred
PUREGRIP Universal HPLC Solvent Reservoir, 38-430 Cap Assembly, 3 x Ports- OD Tube-3.2 mm (1/8") & 1.6 mm (1/16"), 1/EA
Catalog No. 50-255-0697