Learn More
Kentek Corp PUREX FUME EXT HOSE CON KIT
SDP
Supplier:  Kentek Corp PX100032
Encompass
Purex fume extraction hose connection kit for use with 400i, 400i PVC orFumecube Max units. 3 meter length x 50mm diameter
Catalog No. NC3673136