Learn More
Biolegend Purified anti-AMD1
Supplier:  Biolegend 660852
Encompass_Preferred
Purified anti-AMD1 [W18041A]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: WB, IHC-P; Size: 100 μg
Catalog No. 50-286-5977