Learn More
Biolegend Purified anti-β-Tubulin
Supplier:  Biolegend 903401
Encompass_Preferred
Purified anti-β-Tubulin [TU27/Tubulin]; Isotype: Mouse IgG1; Reactivity: Bovine, Rat, Chinese Hamster, Sea Urchin, Chlamydamonas, Drosophila; Apps: WB, ICC, IHC; Size: 250 uL
Catalog No. 50-103-0148