Learn More
Biolegend Purified anti-BRST-2 (GCDFP-15)
Supplier:  Biolegend 915106
Encompass_Preferred
Purified anti-BRST-2 (GCDFP-15) [D6]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: IHC-P; Size: 100 ug
Catalog No. 50-207-5203