Learn More
Biolegend Purified anti-human CD279 PD-1
Supplier:  Biolegend 621602
Encompass_Preferred
Purified anti-human CD279 (PD-1) [A17188B]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: FC, Direct ELISA; Size: 100 μg
Catalog No. 50-272-1807