Learn More
Biolegend Purified anti-human IgG Fc
Supplier:  Biolegend 410701
Encompass_Preferred
Purified anti-human IgG Fc [M1310G05]; Isotype: Rat IgG2a, κ; Reactivity: Human; Apps: FC, ELISA; Size: 100 ug
Catalog No. NC1610074