Learn More
Biolegend Purified anti-Keratin 10
Supplier:  Biolegend 905404
Encompass_Preferred
Purified anti-Keratin 10 [Poly19054]; Isotype: Rabbit Polyclonal; Reactivity: Human, Mouse, Cross-Reactivity: Dog, Primate; Apps: IHC-P, WB; Size: 100 ug
Catalog No. 50-207-5158