Learn More
Biolegend Purified anti-mouse IL-15
Supplier:  Biolegend 631552
Encompass_Preferred
Purified anti-mouse IL-15 [W19387G]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: Direct ELISA; Size: 100 μg
Catalog No. 50-279-6566