Learn More
Biolegend Purified anti-SMAD1 Phospho (S463/S465)/SMAD9 (S465/S467)
Supplier:  Biolegend 610401
Encompass_Preferred
Purified anti-SMAD1 Phospho (S463/S465)/SMAD9 (S465/S467) [A17019A]; Isotype: Mouse IgG1, λ; Reactivity: Mouse, Human; Apps: WB; Size: 25 ug
Catalog No. 50-208-6688