Learn More
Biolegend Purified anti-STK4
Supplier:  Biolegend 673152
Encompass_Preferred
Purified anti-STK4 [W21145C]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: WB, IHC-P; Size: 100 μg
Catalog No. 50-286-6019