Learn More
Biolegend Purified anti-Neurofilament Marker (pan axonal, cocktail)
Supplier:  Biolegend 837904
Encompass_Preferred
Purified anti-Neurofilament Marker (pan axonal, cocktail) [SMI 312]; Isotype: Mouse IgG1/Mouse IgM; Reactivity: Human, Mouse, Rat; Apps: IHC-P, WB, EM, ICC, IHC-F; Size: 100 ug
Catalog No. NC1239357