Learn More
Biolegend Purified azide-free anti-Tau 2
Supplier:  Biolegend 806401
Encompass_Preferred
Purified (azide-free) anti-Tau, 210-230 [Tau 5]; Isotype: Mouse IgG1; Reactivity: Human; Apps: WB, IHC-P; Size: 200 μL
Catalog No. 50-272-1667